siRNA Details
| Gene Symbol: Fes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108488.1 | NP_001101958.1 | 11 | 1602 | ||
| GenBank mRNA | BC166489.1 | - | 11 | 1560 | 7104787 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fes| Gene Symbol: Fes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108488.1 | NP_001101958.1 | 11 | 1602 | ||
| GenBank mRNA | BC166489.1 | - | 11 | 1560 | 7104787 | |