siRNA Details
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108488.1 | NP_001101958.1 | 9 | 1342 | ||
GenBank mRNA | BC166489.1 | - | 9 | 1265 | 7104787 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108488.1 | NP_001101958.1 | 9 | 1342 | ||
GenBank mRNA | BC166489.1 | - | 9 | 1265 | 7104787 |