siRNA Details
| Gene Symbol: Iqgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108489.1 | NP_001101959.2 | 29 | 3819 | ||
| GenBank mRNA | BC169057.1 | - | 1657 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqgap1| Gene Symbol: Iqgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108489.1 | NP_001101959.2 | 29 | 3819 | ||
| GenBank mRNA | BC169057.1 | - | 1657 | |||