siRNA Details
| Gene Symbol: Tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012358.1 | NP_001012358.1 | 4 | 518 | ||
| XM_008759766.1 | XP_008757988.1 | 4 | 639 | |||
| GenBank mRNA | BC089866.1 | - | 4 | 518 | 7321590 | |
| FQ228941.1 | - | 4 | 579 | |||

Tmem41b
Genomic Map