siRNA Details
| Gene Symbol: Tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012358.1 | NP_001012358.1 | 7 | 874 | ||
| GenBank mRNA | BC089866.1 | - | 7 | 874 | 7321590 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem41b| Gene Symbol: Tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012358.1 | NP_001012358.1 | 7 | 874 | ||
| GenBank mRNA | BC089866.1 | - | 7 | 874 | 7321590 | |