siRNA Details
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108499.1 | NP_001101969.1 | 3 | 347 | ||
XM_006230081.2 | XP_006230143.1 | 3 | 1358 | |||
XM_006230082.2 | XP_006230144.1 | 3 | 1358 | |||
XM_006230084.2 | XP_006230146.1 | 3 | 1137 | |||
XM_008759769.1 | XP_008757991.1 | 3 | 1358 | |||
GenBank mRNA | BC168986.1 | - | 2 | 765 |
