siRNA Details
| Gene Symbol: Pik3c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108500.1 | NP_001101970.1 | 5 | 1581 | ||
| XM_008759770.1 | XP_008757992.1 | 4 | 1409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3c2a| Gene Symbol: Pik3c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108500.1 | NP_001101970.1 | 5 | 1581 | ||
| XM_008759770.1 | XP_008757992.1 | 4 | 1409 | |||