siRNA Details
| Gene Symbol: Lcmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199405.2 | NP_955437.1 | 3, 4 | 423 | ||
| XM_006230191.2 | XP_006230253.1 | 3, 4 | 459 | |||
| GenBank mRNA | BC063186.1 | - | 3, 4 | 426 | 6890062 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lcmt1| Gene Symbol: Lcmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199405.2 | NP_955437.1 | 3, 4 | 423 | ||
| XM_006230191.2 | XP_006230253.1 | 3, 4 | 459 | |||
| GenBank mRNA | BC063186.1 | - | 3, 4 | 426 | 6890062 | |