siRNA Details
| Gene Symbol: RGD1310127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035517.1 | NP_001030594.1 | 9 | 1024 | ||
| GenBank mRNA | BC099813.1 | - | 9 | 1024 | 7930179 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310127| Gene Symbol: RGD1310127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035517.1 | NP_001030594.1 | 9 | 1024 | ||
| GenBank mRNA | BC099813.1 | - | 9 | 1024 | 7930179 | |