siRNA Details
| Gene Symbol: Mettl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108504.1 | NP_001101974.1 | 5 | 670 | ||
| XM_006230441.1 | XP_006230503.1 | 5 | 665 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl10| Gene Symbol: Mettl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108504.1 | NP_001101974.1 | 5 | 670 | ||
| XM_006230441.1 | XP_006230503.1 | 5 | 665 | |||