siRNA Details
| Gene Symbol: Ebf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108506.1 | NP_001101976.1 | 8, 9 | 1046 | ||
| XM_006230462.2 | XP_006230524.1 | 7, 8 | 1199 | |||
| XM_006230463.2 | XP_006230525.1 | 7, 8 | 1198 | |||
| XM_006230464.2 | XP_006230526.1 | 7, 8 | 1197 | |||
| XM_006230465.2 | XP_006230527.1 | 7, 8 | 1196 | |||
| XM_008759997.1 | XP_008758219.1 | 7, 8 | 1200 | |||
| XM_008759998.1 | XP_008758220.1 | 7, 8 | 1199 | |||
| XM_008759999.1 | XP_008758221.1 | 7, 8 | 1199 | |||
| XM_008760000.1 | XP_008758222.1 | 7, 8 | 1199 | |||
| XM_008760001.1 | XP_008758223.1 | 7, 8 | 1198 | |||
| XM_008760002.1 | XP_008758224.1 | 7, 8 | 1196 | |||

Ebf3
Genomic Map