siRNA Details
| Gene Symbol: Eps8l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108508.1 | NP_001101978.1 | 3 | 559 | ||
| XM_006230562.1 | XP_006230624.1 | 6 | 689 | |||
| GenBank mRNA | BC166805.1 | - | 6 | 469 | 7939004 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eps8l2| Gene Symbol: Eps8l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108508.1 | NP_001101978.1 | 3 | 559 | ||
| XM_006230562.1 | XP_006230624.1 | 6 | 689 | |||
| GenBank mRNA | BC166805.1 | - | 6 | 469 | 7939004 | |