siRNA Details
| Gene Symbol: Tbc1d10c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081980.1 | NP_001075449.1 | 2 | 150 | ||
| XM_006230840.2 | XP_006230902.1 | 2 | 203 | |||
| GenBank mRNA | BC160813.1 | - | 2 | 161 | 7385853 | |
| FQ226277.1 | - | 2 | 161 | |||

Tbc1d10c
Genomic Map