siRNA Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108515.1 | NP_001101985.1 | 6 | 797 | ||
| XM_006230843.2 | XP_006230905.1 | 6 | 569 | |||
| XM_006230844.2 | XP_006230906.1 | 6 | 1168 | |||
| XM_008760152.1 | XP_008758374.1 | 5 | 817 | |||

Kdm2a
Genomic Map