siRNA Details
| Gene Symbol: Tsga10ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004278.3 | NP_001004278.2 | 3 | 618 | ||
| NM_001270736.1 | NP_001257665.1 | 3 | 618 | |||
| NM_001270737.1 | NP_001257666.1 | 3 | 618 | |||
| XM_006230847.2 | XP_006230909.1 | 3 | 893 | |||
| XM_008760155.1 | XP_008758377.1 | 3 | 903 | |||
| XM_008760156.1 | XP_008758378.1 | 3 | 902 | |||
| XM_008760157.1 | XP_008758379.1 | 3 | 904 | |||
| GenBank mRNA | AY690667.1 | - | 3 | 635 | ||
| BC097254.1 | - | 3 | 599 | 7453704 | ||

Tsga10ip
Genomic Map