siRNA Details
| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230940.2 | XP_006231002.1 | 11 | 1374 | ||
| XM_006230941.2 | XP_006231003.1 | 11 | 1353 | |||
| XM_008760201.1 | XP_008758423.1 | 9 | 1879 | |||
| XM_008760202.1 | XP_008758424.1 | 10 | 1774 | |||
| XM_341993.7 | XP_341994.5 | 12 | 1298 | |||
| GenBank mRNA | FQ222518.1 | - | 3 | 2911 | ||

Snx32
Genomic Map