siRNA Details
| Gene Symbol: Syvn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100739.1 | NP_001094209.1 | 10 | 980 | ||
| XM_006230852.2 | XP_006230914.1 | 10 | 1422 | |||
| GenBank mRNA | BC166727.1 | - | 10 | 980 | 7933989 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syvn1| Gene Symbol: Syvn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100739.1 | NP_001094209.1 | 10 | 980 | ||
| XM_006230852.2 | XP_006230914.1 | 10 | 1422 | |||
| GenBank mRNA | BC166727.1 | - | 10 | 980 | 7933989 | |