siRNA Details
| Gene Symbol: Mta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100740.1 | NP_001094210.1 | 16 | 2055 | ||
| GenBank mRNA | BC099177.1 | - | 3 | 378 | ||
| BC166471.1 | - | 16 | 2055 | 9089183 | ||
| FQ212681.1 | - | 3 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mta2| Gene Symbol: Mta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100740.1 | NP_001094210.1 | 16 | 2055 | ||
| GenBank mRNA | BC099177.1 | - | 3 | 378 | ||
| BC166471.1 | - | 16 | 2055 | 9089183 | ||
| FQ212681.1 | - | 3 | 288 | |||