siRNA Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108520.2 | NP_001101990.1 | 3, 4 | 461 | ||
| GenBank mRNA | BC168974.1 | - | 3, 4 | 461 | 7106310 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Patl1| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108520.2 | NP_001101990.1 | 3, 4 | 461 | ||
| GenBank mRNA | BC168974.1 | - | 3, 4 | 461 | 7106310 | |