siRNA Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108520.2 | NP_001101990.1 | 16 | 2014 | ||
| GenBank mRNA | BC168974.1 | - | 16 | 2014 | 7106310 | |
| FQ214988.1 | - | 2 | 142 | |||
| FQ216743.1 | - | 2 | 142 | |||
| FQ217423.1 | - | 2 | 142 | |||
| FQ217820.1 | - | 2 | 142 | |||
| FQ221257.1 | - | 2 | 142 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172635, AK170918, AK170408, AK085013, AK087559, and others.

Patl1
Genomic Map