siRNA Details
Gene Symbol: Tmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108521.1 | NP_001101991.1 | 4, 5 | 355 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108521.1 | NP_001101991.1 | 4, 5 | 355 |