siRNA Details
| Gene Symbol: Spata6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014165.1 | NP_001014187.1 | 10 | 1038 | ||
| XM_006231237.2 | XP_006231299.1 | 10 | 997 | |||
| XM_006231239.2 | XP_006231301.1 | 10 | 914 | |||
| XM_006231240.2 | XP_006231302.1 | 9 | 884 | |||
| XM_008760342.1 | XP_008758564.1 | 11 | 1067 | |||
| XM_008760343.1 | XP_008758565.1 | 11 | 1068 | |||
| GenBank mRNA | BC079024.1 | - | 10 | 1038 | 7113918 | |

Spata6l
Genomic Map