siRNA Details
| Gene Symbol: Poll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014168.1 | NP_001014190.1 | 8 | 1326 | ||
| XM_006231545.2 | XP_006231607.1 | 7 | 1232 | |||
| GenBank mRNA | BC085841.1 | - | 8 | 1326 | 7135812 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Poll| Gene Symbol: Poll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014168.1 | NP_001014190.1 | 8 | 1326 | ||
| XM_006231545.2 | XP_006231607.1 | 7 | 1232 | |||
| GenBank mRNA | BC085841.1 | - | 8 | 1326 | 7135812 | |