siRNA Details
| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_342068.5 | XP_342069.4 | 5 | 461 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam160b1| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_342068.5 | XP_342069.4 | 5 | 461 | ||