siRNA Details
| Gene Symbol: Abhd16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212531.2 | NP_997696.1 | 3, 4 | 342 | ||
| GenBank mRNA | BC091227.1 | - | 3, 4 | 270 | 7317323 | |
| CK653480.1 | - | 342 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd16a| Gene Symbol: Abhd16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212531.2 | NP_997696.1 | 3, 4 | 342 | ||
| GenBank mRNA | BC091227.1 | - | 3, 4 | 270 | 7317323 | |
| CK653480.1 | - | 342 | ||||