siRNA Details
| Gene Symbol: Ehmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212463.1 | NP_997628.1 | 19, 20 | 2754 | ||
| XM_006255954.2 | XP_006256016.2 | 19, 20 | 2789 | |||
| XM_006255955.2 | XP_006256017.2 | 18, 19 | 2687 | |||
| XM_006255956.2 | XP_006256018.2 | 18, 19 | 2656 | |||
| XM_006255957.1 | XP_006256019.1 | 20, 21 | 2613 | |||
| XM_006255958.1 | XP_006256020.1 | 20, 21 | 2583 | |||
| XM_006255959.1 | XP_006256021.1 | 19, 20 | 2481 | |||

Ehmt2
Genomic Map