siRNA Details
Gene Symbol: Rxrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206849.3 | NP_996731.2 | 9 | 1358 | ||
XM_006255960.2 | XP_006256022.1 | 9 | 1880 | |||
XM_006255961.2 | XP_006256023.1 | 9 | 1373 | |||
GenBank mRNA | BC099776.1 | - | 9 | 1260 | ||
M81766.1 | - | 9 | 1187 |
