siRNA Details
Gene Symbol: Rxrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206849.3 | NP_996731.2 | 5, 6 | 959 | ||
XM_006255960.2 | XP_006256022.1 | 5, 6 | 1469 | |||
XM_006255961.2 | XP_006256023.1 | 5, 6 | 962 | |||
GenBank mRNA | BC099776.1 | - | 5, 6 | 861 |
