siRNA Details
| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025726.1 | NP_001020897.1 | 4, 5 | 390 | ||
| XM_008772734.1 | XP_008770956.1 | 4, 5 | 424 | |||
| GenBank mRNA | BC099089.1 | - | 4, 5 | 390 | 7456350 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srpk1| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025726.1 | NP_001020897.1 | 4, 5 | 390 | ||
| XM_008772734.1 | XP_008770956.1 | 4, 5 | 424 | |||
| GenBank mRNA | BC099089.1 | - | 4, 5 | 390 | 7456350 | |