siRNA Details
| Gene Symbol: Rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025727.1 | NP_001020898.1 | 7 | 1407 | ||
| XM_006256225.2 | XP_006256287.1 | 7 | 1484 | |||
| GenBank mRNA | BC099079.1 | - | 7 | 1407 | 7443362 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf8| Gene Symbol: Rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025727.1 | NP_001020898.1 | 7 | 1407 | ||
| XM_006256225.2 | XP_006256287.1 | 7 | 1484 | |||
| GenBank mRNA | BC099079.1 | - | 7 | 1407 | 7443362 | |