siRNA Details
| Gene Symbol: Zfand3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012175.1 | NP_001012175.1 | 3 | 381 | ||
| XM_008772822.1 | XP_008771044.1 | 3 | 275 | |||
| GenBank mRNA | BC085956.1 | - | 3 | 381 | 7134959 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfand3| Gene Symbol: Zfand3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012175.1 | NP_001012175.1 | 3 | 381 | ||
| XM_008772822.1 | XP_008771044.1 | 3 | 275 | |||
| GenBank mRNA | BC085956.1 | - | 3 | 381 | 7134959 | |