siRNA Details
| Gene Symbol: Specc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039455.1 | NP_001034544.1 | 4 | 517 | ||
| XM_006256339.2 | XP_006256401.1 | 4 | 499 | |||
| GenBank mRNA | AY884296.1 | - | 4 | 517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Specc1l| Gene Symbol: Specc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039455.1 | NP_001034544.1 | 4 | 517 | ||
| XM_006256339.2 | XP_006256401.1 | 4 | 499 | |||
| GenBank mRNA | AY884296.1 | - | 4 | 517 | ||