siRNA Details
| Gene Symbol: Bicc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108531.1 | NP_001102001.1 | 13, 14 | 1830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bicc1| Gene Symbol: Bicc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108531.1 | NP_001102001.1 | 13, 14 | 1830 | ||