siRNA Details
| Gene Symbol: RGD1306739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134576.1 | NP_001128048.1 | 5 | 603 | ||
| XM_006256355.2 | XP_006256417.1 | 5 | 634 | |||
| XM_006256356.2 | XP_006256418.1 | 5 | 609 | |||
| XM_008772887.1 | XP_008771109.1 | 5 | 667 | |||

RGD1306739
Genomic Map