siRNA Details
| Gene Symbol: Hnrnph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108532.1 | NP_001102002.1 | 854 | |||
| XM_006256376.2 | XP_006256438.1 | 8 | 2509 | |||
| XM_006256377.2 | XP_006256439.1 | 8 | 2448 | |||
| XM_006256378.2 | XP_006256440.1 | 8 | 2428 | |||
| XM_006256379.2 | XP_006256441.1 | 8 | 3088 | |||
| XM_006256380.2 | XP_006256442.1 | 7 | 1196 | |||
| GenBank mRNA | FQ215209.1 | - | 8 | 1078 | ||
| Gene Symbol: LOC100910795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751958.3 | XP_003752006.2 | 7 | 1203 | ||
| XM_006256396.2 | XP_006256458.1 | 7 | 1248 | |||

Hnrnph3
Genomic Map