siRNA Details
| Gene Symbol: Sar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007739.1 | NP_001007740.1 | 4 | 344 | ||
| XM_006256450.2 | XP_006256512.1 | 5 | 338 | |||
| XM_006256451.2 | XP_006256513.1 | 4 | 362 | |||
| GenBank mRNA | BC079228.1 | - | 4 | 344 | 7132455 | |
| FQ232554.1 | - | 4 | 298 | |||

Sar1a
Genomic Map