siRNA Details
| Gene Symbol: Fam184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256500.2 | XP_006256562.1 | 1, 2 | 729 | ||
| XM_008773011.1 | XP_008771233.1 | 1, 2 | 729 | |||
| XM_008773012.1 | XP_008771234.1 | 1, 2 | 729 | |||
| XM_008773013.1 | XP_008771235.1 | 1, 2 | 729 | |||
| XM_008773014.1 | XP_008771236.1 | 1, 2 | 729 | |||
| XM_008773015.1 | XP_008771237.1 | 2, 3 | 872 | |||
| XM_008773016.1 | XP_008771238.1 | 2, 3 | 872 | |||

Fam184a
Genomic Map