siRNA Details
| Gene Symbol: Cdc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108538.1 | NP_001102008.1 | 9 | 1232 | ||
| XM_008773000.1 | XP_008771222.1 | 11 | 1202 | |||
| GenBank mRNA | BC166724.1 | - | 11 | 1163 | 7933637 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc40| Gene Symbol: Cdc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108538.1 | NP_001102008.1 | 9 | 1232 | ||
| XM_008773000.1 | XP_008771222.1 | 11 | 1202 | |||
| GenBank mRNA | BC166724.1 | - | 11 | 1163 | 7933637 | |