siRNA Details
| Gene Symbol: Hace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108539.3 | NP_001102009.2 | 2, 3 | 130 | ||
| XM_006256595.2 | XP_006256657.1 | 2, 3 | 383 | |||
| XM_006256597.2 | XP_006256659.1 | 2, 3 | 383 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hace1| Gene Symbol: Hace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108539.3 | NP_001102009.2 | 2, 3 | 130 | ||
| XM_006256595.2 | XP_006256657.1 | 2, 3 | 383 | |||
| XM_006256597.2 | XP_006256659.1 | 2, 3 | 383 | |||