siRNA Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108540.1 | NP_001102010.1 | 1, 2 | 167 | ||
| XM_006231706.2 | XP_006231768.1 | 2, 3 | 1267 | |||
| XM_006231708.2 | XP_006231770.1 | 2, 3 | 696 | |||
| XM_006231709.2 | XP_006231771.1 | 2, 3 | 381 | |||
| XM_006231711.2 | XP_006231773.1 | 2, 3 | 1273 | |||

Rfesd
Genomic Map