siRNA Details
Gene Symbol: Rfesd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108540.1 | NP_001102010.1 | 1 | 93 | ||
XM_006231706.2 | XP_006231768.1 | 2 | 1193 | |||
XM_006231708.2 | XP_006231770.1 | 2 | 622 | |||
XM_006231709.2 | XP_006231771.1 | 2 | 307 | |||
XM_006231711.2 | XP_006231773.1 | 2 | 1199 |
