siRNA Details
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760647.1 | XP_008758869.1 | 12 | 967 | ||
XM_008760649.1 | XP_008758871.1 | 12 | 942 | |||
XM_008760651.1 | XP_008758873.1 | 11 | 876 | |||
XM_008760653.1 | XP_008758875.1 | 11 | 851 | |||
GenBank mRNA | FQ221531.1 | - | 12 | 842 |
