siRNA Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014170.1 | NP_001014192.1 | 11 | 922 | ||
| XM_006232338.2 | XP_006232400.1 | 11 | 954 | |||
| XM_006232339.2 | XP_006232401.1 | 10 | 1078 | |||
| XM_006232341.2 | XP_006232403.1 | 11 | 949 | |||
| XM_006232342.2 | XP_006232404.1 | 10 | 969 | |||
| XM_006232343.2 | XP_006232405.1 | 11 | 1129 | |||
| XM_006232344.2 | XP_006232406.1 | 10 | 921 | |||
| XM_006232345.2 | XP_006232407.1 | 11 | 951 | |||
| XM_006232346.2 | XP_006232408.1 | 10 | 966 | |||
| XM_008760993.1 | XP_008759215.1 | 10 | 1075 | |||
| XM_008760994.1 | XP_008759216.1 | 10 | 1077 | |||
| XM_008760995.1 | XP_008759217.1 | 10 | 1078 | |||
| XM_008760996.1 | XP_008759218.1 | 10 | 1077 | |||
| XM_008760997.1 | XP_008759219.1 | 10 | 1077 | |||
| XM_008760998.1 | XP_008759220.1 | 10 | 1077 | |||
| XM_008760999.1 | XP_008759221.1 | 10 | 1075 | |||
| XM_008761000.1 | XP_008759222.1 | 10 | 1074 | |||
| XM_008761001.1 | XP_008759223.1 | 10 | 1074 | |||
| GenBank mRNA | BC081923.1 | - | 11 | 922 | 7112766 | |

Supt20
Genomic Map