siRNA Details
| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108561.1 | NP_001102031.1 | 10 | 998 | ||
| XM_006233165.1 | XP_006233227.1 | 9 | 882 | |||
| XM_006233166.2 | XP_006233228.1 | 10 | 1222 | |||
| XM_006233167.1 | XP_006233229.1 | 9 | 882 | |||
| XM_008761405.1 | XP_008759627.1 | 10 | 1234 | |||
| XM_008761406.1 | XP_008759628.1 | 10 | 1234 | |||
| GenBank mRNA | BC169126.1 | - | 9 | 862 | ||

Ahcyl1
Genomic Map