siRNA Details
| Gene Symbol: Ndst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191849.1 | NP_001178778.1 | 10 | 2239 | ||
| XM_006233274.2 | XP_006233336.1 | 11 | 2560 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndst4| Gene Symbol: Ndst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191849.1 | NP_001178778.1 | 10 | 2239 | ||
| XM_006233274.2 | XP_006233336.1 | 11 | 2560 | |||