siRNA Details
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191849.1 | NP_001178778.1 | 9 | 2008 | ||
XM_006233274.2 | XP_006233336.1 | 10 | 2329 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191849.1 | NP_001178778.1 | 9 | 2008 | ||
XM_006233274.2 | XP_006233336.1 | 10 | 2329 |