siRNA Details
| Gene Symbol: Lmo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009708.1 | NP_001009708.1 | 643 | |||
| XM_006233438.2 | XP_006233500.1 | 409 | ||||
| GenBank mRNA | BC087700.1 | - | 643 | 7314152 | ||
| FQ233103.1 | - | 486 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmo4| Gene Symbol: Lmo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009708.1 | NP_001009708.1 | 643 | |||
| XM_006233438.2 | XP_006233500.1 | 409 | ||||
| GenBank mRNA | BC087700.1 | - | 643 | 7314152 | ||
| FQ233103.1 | - | 486 | ||||