siRNA Details
| Gene Symbol: Fam73a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761559.1 | XP_008759781.1 | 5 | 607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam73a| Gene Symbol: Fam73a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761559.1 | XP_008759781.1 | 5 | 607 | ||