siRNA Details
| Gene Symbol: Cryz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012183.2 | NP_001012183.1 | 6 | 656 | ||
| XM_006233520.2 | XP_006233582.1 | 7 | 892 | |||
| GenBank mRNA | BC078927.1 | - | 6 | 658 | 7106704 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryz| Gene Symbol: Cryz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012183.2 | NP_001012183.1 | 6 | 656 | ||
| XM_006233520.2 | XP_006233582.1 | 7 | 892 | |||
| GenBank mRNA | BC078927.1 | - | 6 | 658 | 7106704 | |