siRNA Details
| Gene Symbol: Ehmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108572.1 | NP_001102042.1 | 12 | 2087 | ||
| XM_006233618.2 | XP_006233680.2 | 13 | 2203 | |||
| XM_006233619.2 | XP_006233681.2 | 13 | 2194 | |||
| XM_008761598.1 | XP_008759820.1 | 13 | 2545 | |||
| XM_008761599.1 | XP_008759821.1 | 13 | 2542 | |||
| XM_008761600.1 | XP_008759822.1 | 13 | 2524 | |||
| XM_008761601.1 | XP_008759823.1 | 12 | 2401 | |||
| XM_008761602.1 | XP_008759824.1 | 12 | 2380 | |||

Ehmt1
Genomic Map